Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022312.3 | NP_071707.2 | 18 - 19 | 3994 | 69 | |
GenBank mRNA | AK039390.1 | - | 18 - 19 | 3995 | 69 | |
AK078229.1 | - | 11 - 12 | 1672 | 69 | ||
BC132392.1 | - | 18 - 19 | 3628 | 40130669 | 69 | |
BC138043.1 | - | 18 - 19 | 3628 | 8861062 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |