Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127346.1 | NP_001120818.1 | 2 - 3 | 268 | 73 | |
GenBank mRNA | AB183434.1 | - | 2 - 3 | 212 | 73 | |
AK007894.1 | - | 2 - 3 | 259 | 73 | ||
AK166492.1 | - | 2 - 3 | 235 | 73 | ||
AY262012.1 | - | 2 - 3 | 240 | 73 | ||
BC048512.1 | - | 2 - 3 | 227 | 73 | ||
BC116975.1 | - | 2 - 3 | 234 | 40126294 | 73 | |
BC116977.1 | - | 2 - 3 | 234 | 40126296 | 73 | |
BY251841.1 | - | 2 - 3 | 269 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |