Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027432.3 | NP_081708.1 | 4 - 5 | 584 | 67 | |
GenBank mRNA | AK012014.1 | - | 4 - 5 | 575 | 67 | |
AK032897.1 | - | 2 - 3 | 123 | 67 | ||
AK048488.1 | - | 4 - 5 | 607 | 67 | ||
AK050391.1 | - | 4 - 5 | 584 | 67 | ||
AK078856.1 | - | 5 - 6 | 543 | 67 | ||
AK131608.1 | - | 3 - 4 | 495 | 67 | ||
AK161812.1 | - | 4 - 5 | 559 | 67 | ||
AK163839.1 | - | 579 | 67 | |||
AK168006.1 | - | 4 - 5 | 586 | 67 | ||
AK168752.1 | - | 4 - 5 | 606 | 67 | ||
AK169128.1 | - | 4 - 5 | 585 | 67 | ||
BC005755.1 | - | 4 - 5 | 539 | 2654151 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |