Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013367.3 | NP_001013385.3 | 7 - 8 | 1315 | 77 | |
GenBank mRNA | AK081874.1 | - | 1 - 2 | 170 | 77 | |
AK147811.1 | - | 7 - 8 | 1293 | 77 | ||
AK160612.1 | - | 7 - 8 | 1296 | 77 | ||
AY885266.1 | - | 7 - 8 | 1274 | 77 | ||
BC086695.1 | - | 7 - 8 | 1285 | 30691866 | 77 | |
CB245934.1 | - | 2 - 3 | 611 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |