Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025507.2 | NP_079783.2 | 4 - 5 | 453 | 77 | |
GenBank mRNA | AK166163.1 | - | 4 - 5 | 454 | 77 | |
AK167695.1 | - | 4 - 5 | 427 | 77 | ||
AX775977.1 | - | 4 - 5 | 428 | 77 | ||
AX775979.1 | - | 4 - 5 | 428 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |