Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142580.1 | NP_001136052.1 | 2 - 3 | 231 | 84 | |
NM_001142581.1 | NP_001136053.1 | 2 - 3 | 231 | 84 | ||
NM_134044.3 | NP_598805.2 | 2 - 3 | 245 | 84 | ||
GenBank mRNA | AK038165.1 | - | 2 - 3 | 199 | 84 | |
AK046322.1 | - | 2 - 3 | 202 | 84 | ||
AK049643.1 | - | 2 - 3 | 178 | 84 | ||
AK079055.1 | - | 2 - 3 | 178 | 84 | ||
AK082261.1 | - | 2 - 3 | 159 | 84 | ||
AK145043.1 | - | 214 | 84 | |||
BB610654.1 | - | 2 - 3 | 232 | 84 | ||
BC016646.1 | - | 2 - 3 | 154 | 2651233 | 84 | |
BC023716.1 | - | 2 - 3 | 256 | 5346489 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |