Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016927.mm.1 |
NM_001164785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016927.mm.1 |
NM_001164785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016927.mm.1 |
NM_177431.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016927.mm.1 |
TC1500002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016928.mm.1 |
NM_001164785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016928.mm.1 |
NM_001164785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016928.mm.1 |
NM_177431.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016928.mm.1 |
TC1500002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016931.mm.1 |
NM_001164785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016931.mm.1 |
NM_001164785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016931.mm.1 |
NM_177431.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016931.mm.1 |
TC1500002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016929.mm.1 |
NM_001164785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016929.mm.1 |
NM_001164785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016929.mm.1 |
NM_177431.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016929.mm.1 |
TC1500002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016926.mm.1 |
NM_001164785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016926.mm.1 |
NM_001164785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016926.mm.1 |
NM_177431.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016926.mm.1 |
TC1500002109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016930.mm.1 |
NM_001164785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016930.mm.1 |
NM_001164785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016930.mm.1 |
NM_177431.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016930.mm.1 |
TC1500002109.mm.1 |
MTA TC |