Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024141.4 | NP_001019312.1 | 3 - 4 | 474 | 97 | |
GenBank mRNA | AB178899.1 | - | 3 - 4 | 477 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus Probe spans exons |