Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130888.1 | NP_570958.1 | 6 - 7 | 562 | 83 | |
GenBank mRNA | AB178901.1 | - | 7 - 8 | 701 | 83 | |
AJ305317.1 | - | 3 - 4 | 231 | 83 | ||
AJ305318.1 | - | 3 - 4 | 231 | 83 | ||
AY260550.1 | - | 6 - 7 | 562 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |