Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130888.1 | NP_570958.1 | 7 - 8 | 650 | 98 | |
GenBank mRNA | AB178901.1 | - | 8 - 9 | 789 | 98 | |
AJ305317.1 | - | 4 - 5 | 319 | 98 | ||
AJ305318.1 | - | 4 - 5 | 319 | 98 | ||
AY260550.1 | - | 7 - 8 | 650 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |