Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130888.1 | NP_570958.1 | 10 - 11 | 911 | 78 | |
GenBank mRNA | AB178901.1 | - | 11 - 12 | 1050 | 78 | |
AJ305318.1 | - | 7 - 8 | 580 | 78 | ||
AY260550.1 | - | 10 - 11 | 911 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |