Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083114.1 | NP_001076583.1 | 3 - 4 | 266 | 116 | |
NM_001285863.1 | NP_001272792.1 | 3 - 4 | 395 | 116 | ||
NM_001285864.1 | NP_001272793.1 | 2 - 3 | 174 | 116 | ||
NM_146062.3 | NP_666174.2 | 4 - 5 | 473 | 116 | ||
NM_175363.4 | NP_780572.1 | 3 - 4 | 266 | 116 | ||
GenBank mRNA | AK081606.1 | - | 3 - 4 | 259 | 116 | |
AK150904.1 | - | 3 - 4 | 217 | 116 | ||
AK158488.1 | - | 3 - 4 | 396 | 116 | ||
AK160138.1 | - | 3 - 4 | 239 | 116 | ||
AK168443.1 | - | 3 - 4 | 267 | 116 | ||
BC031486.1 | - | 4 - 5 | 426 | 3157250 | 116 | |
BC150854.2 | - | 2 - 3 | 170 | 9087765 | 116 | |
BG145871.1 | - | 4 - 5 | 395 | 116 | ||
BY006733.1 | - | 3 - 4 | 267 | 116 | ||
CF734096.1 | - | 2 - 3 | 154 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |