Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083114.1 | NP_001076583.1 | 6 - 7 | 681 | 100 | |
NM_001285863.1 | NP_001272792.1 | 6 - 7 | 810 | 100 | ||
NM_001285864.1 | NP_001272793.1 | 5 - 6 | 589 | 100 | ||
NM_146062.3 | NP_666174.2 | 8 - 9 | 945 | 100 | ||
NM_175363.4 | NP_780572.1 | 7 - 8 | 738 | 100 | ||
GenBank mRNA | AK081606.1 | - | 7 - 8 | 731 | 100 | |
AK150904.1 | - | 7 - 8 | 689 | 100 | ||
AK158488.1 | - | 6 - 7 | 811 | 100 | ||
AK160138.1 | - | 7 - 8 | 711 | 100 | ||
AK168443.1 | - | 6 - 7 | 683 | 100 | ||
BC031486.1 | - | 8 - 9 | 898 | 3157250 | 100 | |
BC150854.2 | - | 5 - 6 | 585 | 9087765 | 100 | |
CF734096.1 | - | 5 - 6 | 569 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |