Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243837.1 | NP_001230766.1 | 15 - 16 | 2191 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |