Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145475.4 | NP_663450.3 | 2 - 3 | 748 | 69 | |
GenBank mRNA | AB079067.1 | - | 2 - 3 | 255 | 69 | |
AK042077.1 | - | 2 - 3 | 339 | 69 | ||
AK052269.1 | - | 2 - 3 | 514 | 69 | ||
AK169528.1 | - | 2 - 3 | 333 | 69 | ||
BC094253.1 | - | 2 - 3 | 747 | 6400872 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |