Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205353.1 | NP_001192282.1 | 16 - 17 | 1777 | 93 | |
NM_172611.4 | NP_766199.1 | 16 - 17 | 1679 | 93 | ||
GenBank mRNA | AK036832.1 | - | 16 - 17 | 1668 | 93 | |
BC021523.1 | - | 12 - 13 | 1053 | 93 | ||
BC085148.1 | - | 16 - 17 | 1735 | 93 | ||
BC139228.1 | - | 16 - 17 | 1668 | 8862250 | 93 | |
BC139229.1 | - | 16 - 17 | 1668 | 8862251 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |