Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025730.3 | NP_080006.3 | 40 - 41 | 6007 | 75 | |
GenBank mRNA | AK034413.1 | - | 17 - 18 | 2630 | 75 | |
AY792512.1 | - | 40 - 41 | 5950 | 75 | ||
BC034074.1 | - | 6 - 7 | 890 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |