Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172594.2 | NP_766182.2 | 7 - 8 | 899 | 129 | |
GenBank mRNA | AK049530.1 | - | 7 - 8 | 972 | 129 | |
AK087450.1 | - | 7 - 8 | 1213 | 129 | ||
BC020331.1 | - | 7 - 8 | 992 | 129 | ||
BC037686.1 | - | 7 - 8 | 975 | 129 | ||
BC050229.1 | - | 7 - 8 | 928 | 129 | ||
BC057112.1 | - | 7 - 8 | 972 | 6417650 | 129 | |
BC082319.1 | - | 7 - 8 | 962 | 6417162 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |