Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6st | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010560.3 | NP_034690.3 | 1 - 2 | 181 | 62 | |
GenBank mRNA | BC020454.1 | - | 1 - 2 | 135 | 62 | |
BC058679.1 | - | 1 - 2 | 142 | 6834623 | 62 | |
CJ177012.1 | - | 1 - 2 | 183 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |