Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081402.1 | NP_001074871.1 | 4 - 5 | 365 | 63 | |
GenBank mRNA | AK159714.1 | - | 4 - 5 | 347 | 63 | |
AK160933.1 | - | 4 - 5 | 300 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |