Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153144.2 | NP_694784.2 | 12 - 13 | 1841 | 132 | |
GenBank mRNA | AB064543.1 | - | 13 - 14 | 1970 | 132 | |
AY633741.1 | - | 12 - 13 | 1841 | 132 | ||
BC139772.1 | - | 11 - 12 | 1741 | 40111779 | 132 | |
BC145605.1 | - | 12 - 13 | 1869 | 9054171 | 132 | |
BC150801.1 | - | 12 - 13 | 1869 | 9087712 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |