Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153144.2 | NP_694784.2 | 2 - 3 | 294 | 136 | |
GenBank mRNA | AB064543.1 | - | 2 - 3 | 294 | 136 | |
AY633741.1 | - | 2 - 3 | 294 | 136 | ||
BC060738.1 | - | 1 - 2 | 250 | 136 | ||
BC139772.1 | - | 1 - 2 | 200 | 40111779 | 136 | |
BC145605.1 | - | 1 - 2 | 202 | 9054171 | 136 | |
BC150801.1 | - | 1 - 2 | 202 | 9087712 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |