Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177178.3 | NP_796152.2 | 13 - 14 | 1873 | 158 | |
GenBank mRNA | AK036527.1 | - | 13 - 14 | 1895 | 158 | |
AK051777.1 | - | 7 - 8 | 880 | 158 | ||
AK079450.1 | - | 13 - 14 | 1873 | 158 | ||
AK153728.1 | - | 13 - 14 | 1824 | 158 | ||
BC095971.1 | - | 13 - 14 | 1655 | 30108856 | 158 | |
BC120727.1 | - | 13 - 14 | 1845 | 40129650 | 158 | |
BC125421.1 | - | 13 - 14 | 1845 | 40129936 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |