Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177178.3 | NP_796152.2 | 1 - 2 | 169 | 85 | |
GenBank mRNA | AK036527.1 | - | 1 - 2 | 190 | 85 | |
AK079450.1 | - | 1 - 2 | 169 | 85 | ||
AK153728.1 | - | 1 - 2 | 120 | 85 | ||
BC095971.1 | - | 1 - 2 | 86 | 30108856 | 85 | |
BC120727.1 | - | 1 - 2 | 141 | 40129650 | 85 | |
BC125421.1 | - | 1 - 2 | 141 | 40129936 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |