Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145432.3 | NP_663407.2 | 6 - 7 | 885 | 55 | |
GenBank mRNA | AK012216.1 | - | 6 - 7 | 829 | 55 | |
AK132494.1 | - | 6 - 7 | 820 | 55 | ||
AK149059.1 | - | 6 - 7 | 679 | 55 | ||
AK158323.1 | - | 6 - 7 | 1015 | 55 | ||
AK169901.1 | - | 5 - 6 | 2312 | 55 | ||
BC065092.1 | - | 6 - 7 | 811 | 6855410 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |