Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178746.4 | NP_848861.1 | 7 - 8 | 825 | 142 | |
GenBank mRNA | AK043402.1 | - | 7 - 8 | 813 | 142 | |
AK044735.1 | - | 7 - 8 | 815 | 142 | ||
BC052361.1 | - | 7 - 8 | 825 | 6705789 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |