Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030888.4 | NP_112150.1 | 1 - 2 | 209 | 113 | |
GenBank mRNA | AF246265.1 | - | 1 - 2 | 186 | 113 | |
AK029352.1 | - | 1 - 2 | 186 | 113 | ||
AK075867.1 | - | 1 - 2 | 186 | 113 | ||
AK081704.1 | - | 1 - 2 | 186 | 113 | ||
AK128898.1 | - | 1 - 2 | 209 | 113 | ||
BC055047.1 | - | 1 - 2 | 171 | 6486507 | 113 | |
BC055859.1 | - | 1 - 2 | 150 | 4238617 | 113 | |
DQ002396.1 | - | 1 - 2 | 87 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |