Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159598.1 | NP_001153070.1 | 4 - 5 | 1552 | 62 | |
NM_009733.2 | NP_033863.2 | 4 - 5 | 1552 | 62 | ||
GenBank mRNA | AF009011.1 | - | 4 - 5 | 1504 | 62 | |
BC011544.1 | - | 4 - 5 | 1271 | 62 | ||
BC113171.1 | - | 4 - 5 | 1284 | 40061055 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |