Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028063.2 | NP_082339.1 | 3 - 4 | 407 | 108 | |
GenBank mRNA | AK005541.1 | - | 3 - 4 | 407 | 108 | |
AK043532.1 | - | 3 - 4 | 414 | 108 | ||
AK044059.1 | - | 3 - 4 | 414 | 108 | ||
AY349617.1 | - | 3 - 4 | 414 | 108 | ||
BC051548.1 | - | 3 - 4 | 408 | 108 | ||
BC061026.1 | - | 3 - 4 | 394 | 30305575 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |