Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033337.4 | NP_001028509.2 | 2 - 3 | 186 | 76 | |
GenBank mRNA | AK078848.1 | - | 2 - 3 | 147 | 76 | |
AK133636.1 | - | 3 - 4 | 240 | 76 | ||
BC024550.1 | - | 2 - 3 | 136 | 76 | ||
BC030849.1 | - | 2 - 3 | 149 | 76 | ||
BC132621.1 | - | 2 - 3 | 171 | 40130898 | 76 | |
BC132623.1 | - | 2 - 3 | 171 | 40130900 | 76 | |
BY776428.1 | - | 2 - 3 | 187 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |