Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025927.4 | NP_080203.1 | 1 - 2 | 228 | 125 | |
GenBank mRNA | AK011605.1 | - | 1 - 2 | 87 | 125 | |
AK081922.1 | - | 1 - 2 | 89 | 125 | ||
AK157771.1 | - | 1 - 2 | 90 | 125 | ||
AK198473.1 | - | 1 - 2 | 66 | 125 | ||
BC020130.1 | - | 1 - 2 | 73 | 3990098 | 125 | |
BY263659.1 | - | 1 - 2 | 228 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |