Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026396.3 | NP_080672.3 | 1 - 2 | 202 | 141 | |
GenBank mRNA | AK002985.1 | - | 1 - 2 | 89 | 141 | |
AK088313.1 | - | 1 - 2 | 186 | 141 | ||
AK168325.1 | - | 1 - 2 | 175 | 141 | ||
DT927497.1 | - | 1 - 2 | 202 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |