Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500009692.mm.1 |
NM_026396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009692.mm.1 |
NM_026396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009692.mm.1 |
TC1500001202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009699.mm.1 |
NM_026396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009699.mm.1 |
NM_026396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009699.mm.1 |
TC1500001202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009693.mm.1 |
NM_026396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009693.mm.1 |
NM_026396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009693.mm.1 |
TC1500001202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009694.mm.1 |
NM_026396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009694.mm.1 |
NM_026396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009694.mm.1 |
TC1500001202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009695.mm.1 |
NM_026396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009695.mm.1 |
NM_026396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009695.mm.1 |
TC1500001202.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009702.mm.1 |
NM_026396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009702.mm.1 |
NM_026396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009702.mm.1 |
TC1500001202.mm.1 |
MTA TC |