Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011889.2 | NP_036019.2 | 6 - 7 | 777 | 74 | |
GenBank mRNA | AK030668.1 | - | 6 - 7 | 745 | 74 | |
AK046722.1 | - | 6 - 7 | 870 | 74 | ||
AK141915.1 | - | 6 - 7 | 704 | 74 | ||
AK164861.1 | - | 6 - 7 | 874 | 74 | ||
AK169916.1 | - | 6 - 7 | 898 | 74 | ||
AK220520.1 | - | 4 - 5 | 576 | 74 | ||
BC055738.1 | - | 6 - 7 | 706 | 6413697 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |