Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010947.3 | NP_035077.1 | 4 - 5 | 1716 | 63 | |
GenBank mRNA | AF128866.1 | - | 4 - 5 | 1556 | 63 | |
AF149094.1 | - | 4 - 5 | 1352 | 63 | ||
AF152418.1 | - | 4 - 5 | 1471 | 63 | ||
AK031199.1 | - | 4 - 5 | 1861 | 63 | ||
AK083904.1 | - | 4 - 5 | 1450 | 63 | ||
AK141856.1 | - | 4 - 5 | 1627 | 63 | ||
BC053038.1 | - | 4 - 5 | 1808 | 63 | ||
BC094362.1 | - | 4 - 5 | 1716 | 6830466 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |