Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007634.4 | NP_031660.3 | 14 - 15 | 1690 | 95 | |
GenBank mRNA | AK030298.1 | - | 1629 | 95 | ||
AK032961.1 | - | 14 - 15 | 1616 | 95 | ||
AK082485.1 | - | 14 - 15 | 1658 | 95 | ||
AK085183.1 | - | 14 - 15 | 1626 | 95 | ||
AK159594.1 | - | 14 - 15 | 1691 | 95 | ||
AK169263.1 | - | 14 - 15 | 1641 | 95 | ||
AK214285.1 | - | 3 - 4 | 241 | 95 | ||
BC037662.1 | - | 14 - 15 | 1636 | 5353169 | 95 | |
U20612.1 | - | 14 - 15 | 1628 | 95 | ||
U20636.1 | - | 14 - 15 | 1745 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |