Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007634.4 | NP_031660.3 | 3 - 4 | 378 | 78 | |
GenBank mRNA | AK030298.1 | - | 317 | 78 | ||
AK031613.1 | - | 3 - 4 | 318 | 78 | ||
AK032961.1 | - | 3 - 4 | 304 | 78 | ||
AK082485.1 | - | 3 - 4 | 346 | 78 | ||
AK085183.1 | - | 3 - 4 | 315 | 78 | ||
AK159594.1 | - | 3 - 4 | 379 | 78 | ||
AK162447.1 | - | 3 - 4 | 318 | 78 | ||
AK169263.1 | - | 3 - 4 | 329 | 78 | ||
AK211279.1 | - | 2 - 3 | 216 | 78 | ||
BC021658.1 | - | 3 - 4 | 298 | 78 | ||
BC037662.1 | - | 3 - 4 | 330 | 5353169 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |