Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK156579.1 | - | 3 - 4 | 686 | 115 | |
AK218464.1 | - | 2 - 3 | 232 | 115 | ||
BC026523.1 | - | 3 - 4 | 1089 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |