Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024716.1 | NP_001019887.1 | 11 - 12 | 1644 | 79 | |
NM_001039156.1 | NP_001034245.1 | 22 - 23 | 6411 | 79 | ||
NM_138579.4 | NP_613045.3 | 21 - 22 | 6273 | 79 | ||
GenBank mRNA | AK020187.1 | - | 1 - 2 | 76 | 79 | |
AK053194.1 | - | 11 - 12 | 1604 | 79 | ||
AK085512.1 | - | 11 - 12 | 1615 | 79 | ||
AK129417.1 | - | 10 - 11 | 3069 | 79 | ||
AK170390.1 | - | 11 - 12 | 1590 | 79 | ||
BC003984.1 | - | 9 - 10 | 1440 | 79 | ||
BC042760.1 | - | 11 - 12 | 1583 | 79 | ||
BC053329.1 | - | 11 - 12 | 1628 | 79 | ||
BU058377.1 | - | 4 - 5 | 522 | 79 | ||
CA322031.1 | - | 1 - 2 | 64 | 79 | ||
DQ228000.1 | - | 22 - 23 | 6411 | 79 | ||
DQ228001.2 | - | 21 - 22 | 6273 | 79 | ||
DQ278602.1 | - | 20 - 21 | 5895 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |