Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024716.1 | NP_001019887.1 | 9 - 10 | 1364 | 80 | |
NM_001039156.1 | NP_001034245.1 | 20 - 21 | 6131 | 80 | ||
NM_138579.4 | NP_613045.3 | 19 - 20 | 5993 | 80 | ||
GenBank mRNA | AK053194.1 | - | 9 - 10 | 1324 | 80 | |
AK085512.1 | - | 9 - 10 | 1335 | 80 | ||
AK129417.1 | - | 8 - 9 | 2789 | 80 | ||
AK155197.1 | - | 12 - 13 | 3699 | 80 | ||
AK170390.1 | - | 9 - 10 | 1310 | 80 | ||
AK209003.1 | - | 2 - 3 | 138 | 80 | ||
BC003984.1 | - | 7 - 8 | 1160 | 80 | ||
BC042760.1 | - | 9 - 10 | 1303 | 80 | ||
BC053329.1 | - | 9 - 10 | 1348 | 80 | ||
BU058377.1 | - | 2 - 3 | 242 | 80 | ||
DQ228000.1 | - | 20 - 21 | 6131 | 80 | ||
DQ228001.2 | - | 19 - 20 | 5993 | 80 | ||
DQ278602.1 | - | 18 - 19 | 5615 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |