Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161712.1 | NP_001155184.1 | 2 - 3 | 378 | 76 | |
NM_013847.4 | NP_038875.3 | 2 - 3 | 378 | 76 | ||
GenBank mRNA | AF093403.1 | - | 2 - 3 | 336 | 76 | |
AK013138.1 | - | 2 - 3 | 333 | 76 | ||
AK075782.1 | - | 2 - 3 | 318 | 76 | ||
AK076469.1 | - | 2 - 3 | 351 | 76 | ||
AK087433.1 | - | 2 - 3 | 337 | 76 | ||
AK183376.1 | - | 1 - 2 | 153 | 76 | ||
AK187910.1 | - | 2 - 3 | 154 | 76 | ||
BC024107.1 | - | 2 - 3 | 378 | 5125361 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |