Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177461.1 | NP_803412.1 | 12 - 13 | 2562 | 53 | |
GenBank mRNA | AK076763.1 | - | 6 - 7 | 1219 | 53 | |
AK077217.1 | - | 5 - 6 | 1390 | 53 | ||
AK129419.1 | - | 12 - 13 | 2300 | 53 | ||
AK147587.1 | - | 12 - 13 | 2363 | 53 | ||
AK147646.1 | - | 12 - 13 | 2575 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |