Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010444.2 | NP_034574.1 | 5 - 6 | 1494 | 79 | |
GenBank mRNA | AK004709.1 | - | 5 - 6 | 1488 | 79 | |
AK157203.1 | - | 5 - 6 | 1494 | 79 | ||
BC004770.1 | - | 5 - 6 | 1474 | 3156434 | 79 | |
CT010287.1 | - | 4 - 5 | 1366 | 79 | ||
J04113.1 | - | 5 - 6 | 1477 | 79 | ||
X60132.1 | - | 5 - 6 | 1444 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |