Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038658.2 | NP_001033747.1 | 4 - 5 | 919 | 143 | |
NM_028224.4 | NP_082500.2 | 4 - 5 | 955 | 143 | ||
GenBank mRNA | AF468028.1 | - | 3 - 4 | 384 | 143 | |
AK013476.1 | - | 4 - 5 | 358 | 143 | ||
AK030513.1 | - | 4 - 5 | 442 | 143 | ||
AK081252.1 | - | 4 - 5 | 473 | 143 | ||
AK129246.1 | - | 4 - 5 | 468 | 143 | ||
AK136791.1 | - | 4 - 5 | 437 | 143 | ||
AK158960.1 | - | 4 - 5 | 748 | 143 | ||
BC072560.1 | - | 4 - 5 | 955 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |