Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016781.2 | NP_058061.2 | 10 - 11 | 810 | 80 | |
GenBank mRNA | AF036535.1 | - | 10 - 11 | 782 | 80 | |
AK138324.1 | - | 10 - 11 | 776 | 80 | ||
AK159583.1 | - | 10 - 11 | 754 | 80 | ||
BC086660.1 | - | 10 - 11 | 756 | 30551467 | 80 | |
BC109015.1 | - | 6 - 7 | 495 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |