Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Troap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162506.1 | NP_001155978.1 | 7 - 8 | 887 | 118 | |
NM_030159.1 | NP_084435.1 | 7 - 8 | 912 | 118 | ||
GenBank mRNA | AK050381.1 | - | 7 - 8 | 908 | 118 | |
AK077509.1 | - | 7 - 8 | 877 | 118 | ||
BC080746.1 | - | 7 - 8 | 872 | 118 | ||
BC100405.1 | - | 7 - 8 | 841 | 118 | ||
BC145227.1 | - | 7 - 8 | 1285 | 9053779 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |