Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286040.1 | NP_001272969.1 | 20 - 21 | 3115 | 119 | |
NM_001286041.1 | NP_001272970.1 | 19 - 20 | 3052 | 119 | ||
NM_181413.4 | NP_852078.1 | 20 - 21 | 3115 | 119 | ||
GenBank mRNA | AK080878.1 | - | 9 - 10 | 1750 | 119 | |
AK129094.1 | - | 19 - 20 | 2986 | 119 | ||
AK147273.1 | - | 20 - 21 | 3050 | 119 | ||
AK160806.1 | - | 7 - 8 | 789 | 119 | ||
AK169828.1 | - | 9 - 10 | 1061 | 119 | ||
BC010633.1 | - | 8 - 9 | 998 | 119 | ||
BC050847.1 | - | 20 - 21 | 3081 | 6507130 | 119 | |
BC057331.1 | - | 8 - 9 | 1259 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |