Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286040.1 | NP_001272969.1 | 2 - 3 | 420 | 67 | |
NM_001286041.1 | NP_001272970.1 | 2 - 3 | 420 | 67 | ||
NM_181413.4 | NP_852078.1 | 2 - 3 | 420 | 67 | ||
GenBank mRNA | AK129094.1 | - | 2 - 3 | 354 | 67 | |
AK147273.1 | - | 2 - 3 | 355 | 67 | ||
BC027740.1 | - | 2 - 3 | 406 | 67 | ||
BC050847.1 | - | 2 - 3 | 386 | 6507130 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |