Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286041.1 | NP_001272970.1 | 3 - 4 | 578 | 53 | |
GenBank mRNA | AK129094.1 | - | 3 - 4 | 512 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |