Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011950.2 | NP_036080.2 | 9 - 10 | 849 | 94 | |
GenBank mRNA | AK146993.1 | - | 9 - 10 | 837 | 94 | |
U81823.1 | - | 9 - 10 | 806 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |